Assay Details
| Gene Symbol: IL23A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680346.3 | XP_005680403.1 | 2 - 3 | 2765 | 59 | |
| XM_018048070.1 | XP_017903559.1 | 5 - 6 | 1254 | 59 | ||
More Information
Set Membership: |
|

IL23A
Genomic Map