Assay Details
| Gene Symbol: GABRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681572.3 | XP_005681629.1 | 4 - 5 | 722 | 88 | |
| GenBank mRNA | JT734128.1 | - | 6 - 7 | 795 | 81 | |
More Information
Set Membership: |
|

GABRG1
Genomic Map