Assay Details
| Gene Symbol: LOC102175572 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683127.3 | XP_005683184.1 | 4 - 5 | 349 | 80 | |
| XM_005683128.3 | XP_005683185.1 | 3 - 4 | 281 | 80 | ||
| GenBank mRNA | KA348454.1 | - | 3 - 4 | 214 | 80 | |
More Information
Set Membership: |
|

LOC102175572
Genomic Map