Assay Details
| Gene Symbol: DHRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685208.2 | XP_005685265.1 | 8 - 9 | 988 | 62 | |
| XM_005685209.3 | XP_005685266.1 | 8 - 9 | 1106 | 62 | ||
| GenBank mRNA | JO576460.1 | - | 1 - 2 | 454 | 62 | |
More Information
Set Membership: |
|

DHRS1
Genomic Map