Assay Details
| Gene Symbol: OTUD6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689248.3 | XP_005689305.1 | 1 - 2 | 193 | 65 | |
| GenBank mRNA | JO645485.2 | - | 6 - 7 | 901 | 65 | |
More Information
Set Membership: |
|

OTUD6B
Genomic Map