Assay Details
| Gene Symbol: LYPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062512.1 | XP_017918001.1 | 4 - 5 | 646 | 62 | |
| GenBank mRNA | KA327169.1 | - | 4 - 5 | 681 | 62 | |
More Information
Set Membership: |
|

LYPD3
Genomic Map