Assay Details
| Gene Symbol: SLFNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013962863.2 | XP_013818317.2 | 2 - 3 | 2130 | 96 | |
| GenBank mRNA | KA309313.1 | - | 1 - 2 | 571 | 96 | |
More Information
Set Membership: |
|

SLFNL1
Genomic Map