Assay Details
| Gene Symbol: GYG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049119.1 | XP_017904608.1 | 4 - 5 | 685 | 93 | |
| XM_018049124.1 | XP_017904613.1 | 4 - 5 | 597 | 93 | ||
| GenBank mRNA | JO368681.2 | - | 3 - 4 | 1399 | 93 | |
| KA331478.1 | - | 4 - 5 | 595 | 93 | ||
More Information
Set Membership: |
|

GYG1
Genomic Map