Assay Details
| Gene Symbol: CXHXorf38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043941.1 | XP_017899430.1 | 5 - 6 | 878 | 129 | |
| XM_018043942.1 | XP_017899431.1 | 4 - 5 | 745 | 129 | ||
| GenBank mRNA | KA310193.1 | - | 2 - 3 | 266 | 129 | |
More Information
Set Membership: |
|

CXHXorf38
Genomic Map