Assay Details
| Gene Symbol: CXHXorf38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043941.1 | XP_017899430.1 | 6 - 7 | 1041 | 115 | |
| XM_018043942.1 | XP_017899431.1 | 5 - 6 | 908 | 115 | ||
| GenBank mRNA | KA310193.1 | - | 1 - 2 | 103 | 115 | |
More Information
Set Membership: |
|

CXHXorf38
Genomic Map