Assay Details
| Gene Symbol: DNAJC22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679997.3 | XP_005680054.2 | 2 - 3 | 1094 | 77 | |
| GenBank mRNA | JO402764.2 | - | 1 - 2 | 379 | 77 | |
| JO435783.2 | - | 2 - 3 | 1018 | 77 | ||
More Information
Set Membership: |
|

DNAJC22
Genomic Map