Assay Details
| Gene Symbol: CHPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048172.1 | XP_017903661.1 | 8 - 9 | 1371 | 87 | |
| XM_018048173.1 | XP_017903662.1 | 7 - 8 | 1245 | 87 | ||
| XM_018048174.1 | XP_017903663.1 | 7 - 8 | 1239 | 87 | ||
More Information
Set Membership: |
|

CHPT1
Genomic Map