Assay Details
| Gene Symbol: ZFP57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696652.3 | XP_005696709.2 | 1 - 2 | 1371 | 93 | |
| XM_018038814.1 | XP_017894303.1 | 2 - 3 | 606 | 93 | ||
| XM_018038816.1 | XP_017894305.1 | 2 - 3 | 606 | 93 | ||
More Information
Set Membership: |
|

ZFP57
Genomic Map