Assay Details
| Gene Symbol: ZFP57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696652.3 | XP_005696709.2 | 3 - 4 | 1577 | 71 | |
| XM_018038814.1 | XP_017894303.1 | 4 - 5 | 812 | 71 | ||
| XM_018038815.1 | XP_017894304.1 | 4 - 5 | 809 | 71 | ||
More Information
Set Membership: |
|

ZFP57
Genomic Map