Assay Details
| Gene Symbol: RNASEH2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043390.1 | XP_017898879.1 | 2 - 3 | 438 | 92 | |
| XM_018043391.1 | XP_017898880.1 | 2 - 3 | 432 | 92 | ||
| GenBank mRNA | JO636364.2 | - | 3 - 4 | 488 | 92 | |
More Information
Set Membership: |
|

RNASEH2C
Genomic Map