Assay Details
| Gene Symbol: FOSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699905.3 | XP_005699962.2 | 3 - 4 | 487 | 66 | |
| GenBank mRNA | JO346012.2 | - | 3 - 4 | 389 | 66 | |
More Information
Set Membership: |
|

FOSL1
Genomic Map