Assay Details
| Gene Symbol: TAPBPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048594.1 | XP_017904083.1 | 4 - 5 | 1386 | 64 | |
| XM_018048595.1 | XP_017904084.1 | 4 - 5 | 1383 | 64 | ||
| XM_018048596.1 | XP_017904085.1 | 4 - 5 | 1386 | 64 | ||
| XM_018048597.1 | XP_017904086.1 | 4 - 5 | 1386 | 64 | ||
More Information
Set Membership: |
|

TAPBPL
Genomic Map