Assay Details
| Gene Symbol: GRSF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049549.1 | XP_017905038.1 | 6 - 7 | 1241 | 71 | |
| XM_018049550.1 | XP_017905039.1 | 6 - 7 | 921 | 71 | ||
More Information
Set Membership: |
|

GRSF1
Genomic Map