Assay Details
| Gene Symbol: CTDSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059502.1 | XP_017914991.1 | 6 - 7 | 2003 | 68 | |
| XM_018059509.1 | XP_017914998.1 | 6 - 7 | 2003 | 68 | ||
| GenBank mRNA | JO637725.2 | - | 6 - 7 | 895 | 68 | |
More Information
Set Membership: |
|

CTDSP1
Genomic Map