Assay Details
| Gene Symbol: ADTRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013974157.2 | XP_013829611.2 | 5 - 6 | 936 | 62 | |
| GenBank mRNA | JO421525.2 | - | 5 - 6 | 1119 | 62 | |
More Information
Set Membership: |
|

ADTRP
Genomic Map