Assay Details
| Gene Symbol: RELCH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039531.1 | XP_017895020.1 | 9 - 10 | 1777 | 65 | |
| XM_018039532.1 | XP_017895021.1 | 9 - 10 | 1737 | 65 | ||
| XM_018039533.1 | XP_017895022.1 | 8 - 9 | 1441 | 65 | ||
| XM_018039534.1 | XP_017895023.1 | 9 - 10 | 1777 | 65 | ||
More Information
Set Membership: |
|

RELCH
Genomic Map