Assay Details
| Gene Symbol: CALU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679443.3 | XP_005679500.1 | 5 - 6 | 771 | 79 | |
| XM_005679444.3 | XP_005679501.1 | 5 - 6 | 772 | 79 | ||
| GenBank mRNA | JR134268.1 | - | 2306 | 79 | ||
More Information
Set Membership: |
|

CALU
Genomic Map