Assay Details
| Gene Symbol: LOC102174373 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681282.3 | XP_005681339.2 | 11 - 12 | 2966 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102174373| Gene Symbol: LOC102174373 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681282.3 | XP_005681339.2 | 11 - 12 | 2966 | 76 | |
Set Membership: |
|