Assay Details
| Gene Symbol: RNF145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050095.1 | XP_017905584.1 | 6 - 7 | 1003 | 64 | |
| XM_018050096.1 | XP_017905585.1 | 5 - 6 | 975 | 64 | ||
| XM_018050097.1 | XP_017905586.1 | 7 - 8 | 1067 | 64 | ||
More Information
Set Membership: |
|

RNF145
Genomic Map