Assay Details
| Gene Symbol: FMO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690637.3 | XP_005690694.3 | 5 - 6 | 971 | 73 | |
| XM_013970175.2 | XP_013825629.1 | 5 - 6 | 971 | 73 | ||
| GenBank mRNA | JO427134.1 | - | 5 - 6 | 1034 | 73 | |
More Information
Set Membership: |
|

FMO4
Genomic Map