Assay Details
| Gene Symbol: PPP1R14A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062325.1 | XP_017917814.1 | 3 - 4 | 576 | 78 | |
| GenBank mRNA | JO563016.2 | - | 3 - 4 | 432 | 78 | |
| JO579792.1 | - | 3 - 4 | 426 | 78 | ||
| JO634842.2 | - | 1 - 2 | 686 | 78 | ||
More Information
Set Membership: |
|

PPP1R14A
Genomic Map