Assay Details
| Gene Symbol: SNTB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058133.1 | XP_017913622.1 | 5 - 6 | 1476 | 60 | |
| GenBank mRNA | JT697569.1 | - | 3 - 4 | 317 | 60 | |
More Information
Set Membership: |
|

SNTB1
Genomic Map