Assay Details
| Gene Symbol: METTL24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018053074.1 | XP_017908563.1 | 2 - 3 | 503 | 66 | |
| GenBank mRNA | JT661365.1 | - | 3 - 4 | 289 | 66 | |
More Information
Set Membership: |
|

METTL24
Genomic Map