Assay Details
| Gene Symbol: FAM3D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013973520.2 | XP_013828974.1 | 4 - 5 | 326 | 56 | |
| XM_018038466.1 | XP_017893955.1 | 4 - 5 | 384 | 56 | ||
More Information
Set Membership: |
|

FAM3D
Genomic Map