Assay Details
| Gene Symbol: UNC5CL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039294.1 | XP_017894783.1 | 2 - 3 | 722 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UNC5CL| Gene Symbol: UNC5CL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039294.1 | XP_017894783.1 | 2 - 3 | 722 | 59 | |
Set Membership: |
|