Assay Details
| Gene Symbol: ANKRD23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054869.1 | XP_017910358.1 | 5 - 6 | 595 | 55 | |
| GenBank mRNA | JO344383.2 | - | 4 - 5 | 1546 | 55 | |
More Information
Set Membership: |
|

ANKRD23
Genomic Map