Assay Details
| Gene Symbol: TMEM41B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689709.2 | XP_005689766.1 | 6 - 7 | 966 | 93 | |
| GenBank mRNA | JT697972.1 | - | 6 - 7 | 887 | 93 | |
More Information
Set Membership: |
|

TMEM41B
Genomic Map