Assay Details
| Gene Symbol: CNST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690548.3 | XP_005690605.1 | 5 - 6 | 843 | 77 | |
| XM_018060245.1 | XP_017915734.1 | 5 - 6 | 862 | 77 | ||
| XM_018060246.1 | XP_017915735.1 | 6 - 7 | 975 | 77 | ||
| GenBank mRNA | JT708951.1 | - | 9 - 10 | 1691 | 77 | |
More Information
Set Membership: |
|

CNST
Genomic Map