Assay Details
| Gene Symbol: CFHR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691048.3 | XP_005691105.2 | 9 - 10 | 1598 | 71 | |
| XM_013970400.2 | XP_013825854.2 | 9 - 10 | 1571 | 71 | ||
More Information
Set Membership: |
|

CFHR5
Genomic Map