Assay Details
| Gene Symbol: ZNF827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061580.1 | XP_017917069.1 | 2 - 3 | 1687 | 75 | |
| XM_018061581.1 | XP_017917070.1 | 2 - 3 | 475 | 75 | ||
| XM_018061582.1 | XP_017917071.1 | 2 - 3 | 1687 | 75 | ||
More Information
Set Membership: |
|

ZNF827
Genomic Map