Assay Details
| Gene Symbol: APOBEC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696288.3 | XP_005696345.2 | 2 - 3 | 852 | 66 | |
| GenBank mRNA | JO366318.2 | - | 2 - 3 | 892 | 66 | |
More Information
Set Membership: |
|

APOBEC2
Genomic Map