Assay Details
| Gene Symbol: AP3M1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699225.3 | XP_005699282.1 | 7 - 8 | 1375 | 68 | |
| XM_005699226.3 | XP_005699283.1 | 7 - 8 | 1404 | 68 | ||
More Information
Set Membership: |
|

AP3M1
Genomic Map