Assay Details
| Gene Symbol: WDR34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055903.1 | XP_017911392.1 | 8 - 9 | 1424 | 60 | |
| XM_018055904.1 | XP_017911393.1 | 8 - 9 | 1377 | 60 | ||
| XM_018055905.1 | XP_017911394.1 | 7 - 8 | 2007 | 60 | ||
| XM_018055906.1 | XP_017911395.1 | 8 - 9 | 1379 | 60 | ||
| GenBank mRNA | JO432870.2 | - | 1 - 2 | 306 | 60 | |
| KA307757.1 | - | 1 - 2 | 300 | 60 | ||
More Information
Set Membership: |
|

WDR34
Genomic Map