Assay Details
| Gene Symbol: BLVRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062427.1 | XP_017917916.1 | 4 - 5 | 569 | 76 | |
| GenBank mRNA | JO323129.2 | - | 1 - 2 | 272 | 76 | |
More Information
Set Membership: |
|

BLVRB
Genomic Map