Assay Details
| Gene Symbol: SLC16A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065407.1 | XP_017920896.1 | 3 - 4 | 480 | 63 | |
| XM_018065408.1 | XP_017920897.1 | 3 - 4 | 542 | 63 | ||
| XM_018065409.1 | XP_017920898.1 | 2 - 3 | 352 | 63 | ||
| GenBank mRNA | JO427251.1 | - | 3 - 4 | 2050 | 63 | |
More Information
Set Membership: |
|

SLC16A3
Genomic Map