Assay Details
| Gene Symbol: LOC102184887 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013976215.2 | XP_013831669.1 | 15 - 16 | 4077 | 109 | |
| XM_013976216.2 | XP_013831670.1 | 15 - 16 | 4077 | 109 | ||
| XM_013976217.2 | XP_013831671.1 | 15 - 16 | 4077 | 109 | ||
| XM_013976218.2 | XP_013831672.1 | 15 - 16 | 4077 | 109 | ||
| XM_013976219.2 | XP_013831673.1 | 15 - 16 | 1576 | 109 | ||
| XM_013976220.2 | XP_013831674.1 | 15 - 16 | 1550 | 109 | ||
More Information
Set Membership: |
|

LOC102184887
Genomic Map