Assay Details
| Gene Symbol: LOC102184887 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013976215.2 | XP_013831669.1 | 1 - 2 | 2588 | 74 | |
| XM_013976216.2 | XP_013831670.1 | 1 - 2 | 2588 | 74 | ||
| XM_013976217.2 | XP_013831671.1 | 1 - 2 | 2588 | 74 | ||
| XM_013976218.2 | XP_013831672.1 | 1 - 2 | 2588 | 74 | ||
More Information
Set Membership: |
|

LOC102184887
Genomic Map