Assay Details
| Gene Symbol: LOC102184887 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013976215.2 | XP_013831669.1 | 20 - 21 | 4529 | 177 | |
| XM_013976216.2 | XP_013831670.1 | 20 - 21 | 4520 | 177 | ||
| XM_013976217.2 | XP_013831671.1 | 19 - 20 | 4457 | 177 | ||
| XM_013976219.2 | XP_013831673.1 | 20 - 21 | 2028 | 177 | ||
| XM_013976220.2 | XP_013831674.1 | 20 - 21 | 2002 | 177 | ||
More Information
Set Membership: |
|

LOC102184887
Genomic Map