Assay Details
| Gene Symbol: LOC102184887 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013976215.2 | XP_013831669.1 | 2 - 3 | 2676 | 96 | |
| XM_013976216.2 | XP_013831670.1 | 2 - 3 | 2676 | 96 | ||
| XM_013976217.2 | XP_013831671.1 | 2 - 3 | 2676 | 96 | ||
| XM_013976218.2 | XP_013831672.1 | 2 - 3 | 2676 | 96 | ||
| XM_013976219.2 | XP_013831673.1 | 2 - 3 | 175 | 96 | ||
| XM_013976220.2 | XP_013831674.1 | 2 - 3 | 149 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013976219.2, XM_013976220.2 transcripts.
Set Membership: |
|

LOC102184887
Genomic Map