Assay Details
| Gene Symbol: LOC102184887 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013976215.2 | XP_013831669.1 | 7 - 8 | 3094 | 88 | |
| XM_013976216.2 | XP_013831670.1 | 7 - 8 | 3094 | 88 | ||
| XM_013976217.2 | XP_013831671.1 | 7 - 8 | 3094 | 88 | ||
| XM_013976218.2 | XP_013831672.1 | 7 - 8 | 3094 | 88 | ||
| XM_013976219.2 | XP_013831673.1 | 7 - 8 | 593 | 88 | ||
| XM_013976220.2 | XP_013831674.1 | 7 - 8 | 567 | 88 | ||
| GenBank mRNA | JO452221.2 | - | 3 - 4 | 207 | 88 | |
More Information
Set Membership: |
|

LOC102184887
Genomic Map