Assay Details
| Gene Symbol: FRZB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675886.3 | XP_005675943.1 | 6 - 7 | 1232 | 67 | |
| XM_005675889.3 | XP_005675946.1 | 6 - 7 | 1278 | 67 | ||
| XM_013968455.2 | XP_013823909.1 | 6 - 7 | 1290 | 67 | ||
| XM_018065459.1 | XP_017920948.1 | 6 - 7 | 1287 | 67 | ||
| GenBank mRNA | JT677341.1 | - | 9 - 10 | 1417 | 67 | |
More Information
Set Membership: |
|

FRZB
Genomic Map