Assay Details
| Gene Symbol: ASB15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679407.2 | XP_005679464.1 | 7 - 8 | 879 | 79 | |
| GenBank mRNA | JO392664.2 | - | 4 - 5 | 1060 | 79 | |
More Information
Set Membership: |
|

ASB15
Genomic Map