Assay Details
| Gene Symbol: SLC26A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683160.3 | XP_005683217.2 | 4 - 5 | 1185 | 71 | |
| XM_005683161.3 | XP_005683218.2 | 5 - 6 | 1364 | 71 | ||
| XM_005683163.3 | XP_005683220.2 | 3 - 4 | 1120 | 71 | ||
| XM_013965688.2 | XP_013821142.2 | 5 - 6 | 1321 | 71 | ||
| XM_013965689.2 | XP_013821143.2 | 4 - 5 | 1081 | 71 | ||
| XM_018050166.1 | XP_017905655.1 | 5 - 6 | 1593 | 71 | ||
| XM_018050167.1 | XP_017905656.1 | 3 - 4 | 1392 | 71 | ||
| GenBank mRNA | JO435380.2 | - | 1 - 2 | 384 | 71 | |
More Information
Set Membership: |
|

SLC26A2
Genomic Map