Assay Details
| Gene Symbol: WIPF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693774.3 | XP_005693831.1 | 4 - 5 | 487 | 74 | |
| XM_018065026.1 | XP_017920515.1 | 4 - 5 | 548 | 74 | ||
| XM_018065027.1 | XP_017920516.1 | 6 - 7 | 934 | 74 | ||
| XM_018065028.1 | XP_017920517.1 | 5 - 6 | 672 | 74 | ||
| XM_018065029.1 | XP_017920518.1 | 4 - 5 | 511 | 74 | ||
| GenBank mRNA | JO446595.2 | - | 1 - 2 | 272 | 74 | |
| KA315274.1 | - | 4 - 5 | 4852 | 74 | ||
More Information
Set Membership: |
|

WIPF2
Genomic Map