Assay Details
| Gene Symbol: CDCP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038392.1 | XP_017893881.1 | 7 - 8 | 2103 | 60 | |
| GenBank mRNA | KA305531.1 | - | 2 - 3 | 915 | 60 | |
More Information
Set Membership: |
|

CDCP1
Genomic Map